Hand-Operated Drum Pumps, HDPE
Supplier: Avantor Fluid Handling
06425-05
MFLX06425-05EA 307 EUR
MFLX06425-05
Hand-Operated Drum Pumps, HDPE
Pumps Drum/Transfer Pumps
Ideal for most acid/base or ionic salt applications.
  • Pump acids, alcohol, and alkalies from carboys, drums, or tanks
  • Self-cleaning ball valve ensures free flow and long pump life
This plastic hand-operated drum pump is ideal for pumping acids, alcohols, and alkalis. Constructed of all HDPE parts, this pump offers 6 strokes/gallon (2 strokes/liter). Order optional LDPE adapter to securely mount pump on drums. Adapter 06425-05 sold separately.