Masterflex® L/S® High-Performance Precision Pump Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
MFLX06508-15EA 193 EUR
MFLX06508-15 MFLX06508-24 MFLX06508-35 MFLX06508-36
Masterflex® L/S® High-Performance Precision Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Protect your light-sensitive samples.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly
More Product Information