Masterflex® Ismatec® Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Supplier: Avantor Fluid Handling
MFLX06512-04EA 90.5 EUR
MFLX06512-04
Masterflex® Ismatec® Pump Tubing, Tygon® Fuel and Lubricant, Avantor®
Tubing Peristaltic Pump Tubing
No more tubing swell when pumping fuel.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Resists most hydrocarbons and embrittlement
This tubing is ideal for use with fuels and industrial lubricants such as gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. ThisMore Product Information