Pump Motor Spider Inserts, Buna-N®
Supplier: Avantor Fluid Handling
MFLX07127-69EA 17.5 EUR
MFLX07127-69
Pump Motor Spider Inserts, Buna-N®
Pumps Motors
Create your own motor mounting with the hardware below.
  • Constructed of durable Buna-N®
Spider smooths power transfer, absorbs vibration, and compensates for minor misalignment.