Masterflex® Ismatec® Microbore Pump Tubing, Peroxide-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
MFLX07625-12EA 171 EUR
MFLX07625-12 MFLX07625-18 MFLX07625-22 MFLX07625-24 MFLX07625-26 MFLX07625-28 MFLX07625-30 MFLX07625-32 MFLX07625-34 MFLX07625-36 MFLX07625-38 MFLX07625-40 MFLX07625-42 MFLX07625-44 MFLX07625-46 MFLX07625-48 MFLX07625-14
Masterflex® Ismatec® Microbore Pump Tubing, Peroxide-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: Demanding high-pressure pharmaceutical, biotechnology, aerospace, and food and beverage applications.
Note: For customMore Product Information