PrimeLock® High-Purity PFA Fittings, NPT(M) Straight Adapters
Supplier: Avantor Fluid Handling
30492-32
MFLX30492-32EA 116 EUR
MFLX30492-32
PrimeLock® High-Purity PFA Fittings, NPT(M) Straight Adapters
Fittings and Connectors
Ultra-clean and reliable fittings for highly critical and corrosive chemicals.
  • Broad chemical compatibility with PFA wetted components
  • Three nonwetted backup seals prevent leaks without introducing sources of entrapment or flow interruptions
  • Audible and visual checks guarantee proper connections
  • Heat not required for fitting installation
PFA wetted fittings handle critical chemical applications for manufacturers and laboratories. Tubing connections offer a primary seal and multiple safety backups with nonwetted inserts, reducing entrapment, leakMore Product Information