CPC® Metal Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Supplier: Avantor Fluid Handling
31403-72 31403-72BLK 31308-91BLK 31308-87BLK 31309-31BLK
MFLX31403-72EA 60 EUR
MFLX31403-72 MFLX31403-72BLK MFLX31308-91BLK MFLX31308-87BLK MFLX31309-31BLK
CPC® Metal Quick-Disconnect Fittings, Panel-Mount Hose Barb Bodies
Fittings and Connectors
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from −40 toMore Product Information