Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Panel-Mount, 65-mm, Avantor®
Supplier: Avantor Fluid Handling
32003-09 32004-09 32005-09 32013-01 32013-16 32013-18 32013-21 32013-24 32013-28 32013-29 32013-31 32013-35 32014-01 32014-07 32014-11 32014-15 32014-16 32014-21 32014-28 32014-29 32015-24 32015-33
MFLX32013-03EA 153 EUR
MFLX32013-03 MFLX32013-05 MFLX32013-07 MFLX32013-09 MFLX32013-11 MFLX32013-13 MFLX32013-15 MFLX32013-17 MFLX32013-19 MFLX32013-27 MFLX32013-33 MFLX32014-03 MFLX32014-05 MFLX32014-09 MFLX32014-13 MFLX32014-17 MFLX32014-24 MFLX32015-01 MFLX32015-03 MFLX32015-05 MFLX32015-07 MFLX32015-09 MFLX32015-11 MFLX32015-13 MFLX32015-15 MFLX32015-16 MFLX32015-17 MFLX32015-19 MFLX32015-21 MFLX32015-23 MFLX32015-29 MFLX32015-31 MFLX32015-35 MFLX32003-09 MFLX32004-09 MFLX32005-09 MFLX32013-01 MFLX32013-16 MFLX32013-18 MFLX32013-21 MFLX32013-24 MFLX32013-28 MFLX32013-29 MFLX32013-31 MFLX32013-35 MFLX32014-01 MFLX32014-07 MFLX32014-11 MFLX32014-15 MFLX32014-16 MFLX32014-21 MFLX32014-28 MFLX32014-29 MFLX32015-24 MFLX32015-33
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, Panel-Mount, 65-mm, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information