Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Aluminum, Avantor®
Supplier: Avantor Fluid Handling
32060-02 32061-00 32061-11 32061-12 32061-14 32062-12 32062-14 32062-30 32062-32
MFLX32060-02EA 427 EUR
MFLX32060-02 MFLX32061-00 MFLX32061-11 MFLX32061-12 MFLX32061-14 MFLX32062-12 MFLX32062-14 MFLX32062-30 MFLX32062-32
Masterflex® Direct-Read Variable-Area Flowmeters for Water, Dual Scale, Aluminum, Avantor®
Flow Meters
Interchangeable medias and scales offer flexibility.
  • Select water or air scale in either English or metric units by rotating the flowtube
  • Dual water scale shows flow rate in gallons per minute and liters per minute
  • Dual air scale shows flow rate in standard cubic feet per minute and liters per minute
The heavy-walled flowtube construction and protective metal housing make these dual media/scale flowmeters perfect for busy industrial areas. Choose from models with a valve to monitor/control flow or without a valve to just monitor flow. Flowmeters areMore Product Information