Masterflex® Direct-Read Variable-Area Flowmeters without Valve, Metric Units, Avantor®
Supplier: Avantor Fluid Handling
32457-32 32457-34 32457-48 32458-34 32462-24
MFLX32457-40EA 65.7 EUR
MFLX32457-40 MFLX32457-42 MFLX32457-44 MFLX32457-46 MFLX32457-50 MFLX32457-52 MFLX32458-30 MFLX32458-32 MFLX32458-36 MFLX32458-38 MFLX32458-40 MFLX32458-42 MFLX32458-44 MFLX32458-46 MFLX32458-50 MFLX32458-52 MFLX32458-54 MFLX32458-56 MFLX32458-58 MFLX32458-60 MFLX32458-62 MFLX32458-64 MFLX32458-65 MFLX32462-20 MFLX32462-30 MFLX32462-32 MFLX32462-34 MFLX32457-32 MFLX32457-34 MFLX32457-48 MFLX32458-34 MFLX32462-24
Masterflex® Direct-Read Variable-Area Flowmeters without Valve, Metric Units, Avantor®
Flow Meters
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is aMore Product Information