Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
Supplier: Avantor Fluid Handling
MFLX74000-49-EUEA 11680 EUR
MFLX74000-49-EU MFLX74000-51-EU MFLX74000-53-EU MFLX74000-59-EU MFLX74000-49-UK MFLX74000-51-UK MFLX74000-53-UK MFLX74000-59-UK
Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
Pumps Diaphragm Pumps
Monitor transfer of delicate biological fluids with EtherNet/IP™ control capability.
  • Choose from readily available single-use or multiple-use Quattroflow™ pump head systems
  • Advanced network and communication models feature EtherNet/IP™
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Ideal for cross-flow systems, tangential flow filtration (TFF), chromatography, cultures, and more
The Quattroflow™More Product Information