Warren Rupp Air-Powered Double-Diaphragm Polypropylene Pumps
Supplier: Avantor Fluid Handling
74000-00 74000-11 74000-13 74000-42 74000-70 74000-72
MFLX74000-00EA 1110 EUR
MFLX74000-00 MFLX74000-11 MFLX74000-13 MFLX74000-42 MFLX74000-70 MFLX74000-72
Warren Rupp Air-Powered Double-Diaphragm Polypropylene Pumps
Pumps Air-Operated Pumps
Move fluids with viscosities up to 11,000 cp.
  • Air-powered motor eliminates sparking concerns associate with electrical pumps
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing
  • Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids
  • Safely handles abrasive fluids and slurries — ideal for high-viscosity and shear sensitive materials
  • Encapsulated muffler provides excellent sound dampening
These pumps can moveMore Product Information