Masterflex® Console Gear Pump Drive for MicroPump A-Mount Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
75211-12
MFLX75211-12EA 1190 EUR
MFLX75211-12 MFLX75211-17-EU MFLX75211-17-UK
Masterflex® Console Gear Pump Drive for MicroPump A-Mount Pump Heads, Avantor®
Pumps Pump Drives Gear Pump Drives
Simple variable speed control handles many fluid handling applications.
  • 100:1 speed-control range controls flow over a wide range
  • Maximum continuous-duty flow rates possible with Micropump® A-mount pump heads
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Remote control of start/stop via DB9 connector on back of drive
Compact, cost-effective drive offers simple variable speed control for many fluid handling applications fromMore Product Information