Masterflex® L/S® Rigid PTFE-Tubing Pump Head, Avantor®
Supplier: Avantor Fluid Handling
MFLX77390-00EA 1160 EUR
MFLX77390-00
Masterflex® L/S® Rigid PTFE-Tubing Pump Head, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for high-purity fluids and aggressive organic solvents.
  • PTFE tubing is the only wetted part ensuring fluid purity and excellent chemical compatibility
  • Delivers a wide range of flow rates at pressures up to 100 psi (6.9 bar)
  • Six-roller pump head with adjustable occlusion—tighten occlusion for higher pressure, loosen for longer tubing life
The PTFE tubing pump head is ideal for applications such as filtration, chemical injection, or high-purity sampling. It is compatible with L/S drives that accept two or more pump heads with a 1/10 hp minimum.More Product Information