Masterflex® Transfer Tubing, Platinum-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
MFLX95802-00EA 145 EUR
MFLX95802-00 MFLX95802-01 MFLX95802-02 MFLX95802-03 MFLX95802-04 MFLX95802-05 MFLX95802-06 MFLX95802-07 MFLX95802-08 MFLX95802-09 MFLX95802-10 MFLX95802-11 MFLX95802-12 MFLX95802-13 MFLX95802-14 MFLX95802-15 MFLX95802-16 MFLX95802-17 MFLX95802-18 MFLX95802-19 MFLX95802-20 MFLX95802-21 MFLX95802-22 MFLX95802-23 MFLX95802-24 MFLX95802-25 MFLX95802-26 MFLX95803-08
Masterflex® Transfer Tubing, Platinum-Cured Silicone, Avantor®
Tubing Transfer Tubing
No need to worry about the heat - use for applications up to 460 °F.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications.
Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless,More Product Information