Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, FDA-Compliant Viton®, Avantor®
Supplier: Avantor Fluid Handling
MFLX97628-18EA 186 EUR
MFLX97628-18 MFLX97628-22 MFLX97628-24 MFLX97628-26 MFLX97628-28 MFLX97628-30 MFLX97628-32 MFLX97628-34 MFLX97628-36 MFLX97628-38 MFLX97628-40 MFLX97628-42 MFLX97628-44 MFLX97628-46 MFLX97628-48 MFLX96428-26 MFLX96428-42 MFLX96428-48 MFLX96428-34
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, FDA-Compliant Viton®, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Acid and solvent transfer; high-temperature work
Characteristics: Opaque black
Resin certification: Tubing is manufactured according to GMP
Temperature range: -8 to 400 °F (-13 to 204More Product Information