Masterflex® Low-Flow Direct-Lift Proportioning Solenoid Valves, Avantor®
Supplier: Avantor Fluid Handling
98650-02 98650-12 98650-32 98650-34 98650-22
MFLX98650-02EA 323 EUR
MFLX98650-02 MFLX98650-12 MFLX98650-32 MFLX98650-34 MFLX98650-22
Masterflex® Low-Flow Direct-Lift Proportioning Solenoid Valves, Avantor®
Valves Solenoid Valves
Resist pressures up to 500 psi!
  • Use for continuous control of liquid or gas flow
  • Valves withstand pressures to 500 psi
Apply a 0 to 30 VDC input signal—these normally-closed valves open to give you flow rates that are proportional to the input voltage. Stainless steel valve construction withstands pressures to 500 psi. Maximum fluid temperature is 122 °F (50 °C). Maximum ambient temperature with intermittent use is 130 °F (54 °C). Actuation time is 500 milliseconds.
Wetted parts are 316 and 416 stainless steel and VITON®. Ports are 1/4" compressionMore Product Information