Bottle brush
Supplier: MICRONOVA
BRWH-67
MICRBRWH-67EA 117 EUR
MICRBRWH-67
Bottle brush
Brushes
This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.
  • BPA-free
Base resins used to produce block and bristles are made from FDA approved materials.