Cleanroom pens
Supplier: MICRONOVA
PEN-01IR PEN-02IR PEN-20IR PEN-02 PEN-01_U PEN-01TIR PEN-25IR PEN-22IR PEN-51IR PEN-11 PEN-11IR PEN-12 PEN-12IR PEN-23IR PEN-51 PEN-52 PEN-52IR PEN-53IR
818-0550EA 66 EUR
818-0550 818-0551 811-0122 818-0244 818-0243 811-0168 811-0174 811-0173 811-0195 811-0192 818-0284 811-0193 818-0285 MICRPEN-23IR MICRPEN-51 MICRPEN-52 811-0196 MICRPEN-53IR
Cleanroom pens
Pens Cleanroom Pens
Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom irradiated Sharpie® markers write permanently on most surfaces with quick drying, non toxic, alcohol-based ink that is fade and water resistant to further extend multiple project uses. Marks will not smear on reusable glass, metal, or porcelain lab ware, but can be scrubbed off.
  • Indelible, low sodium ink pens
  • White, high density polypropylene barrel
  • Holds up to wipe down with IPA, bleach, and phenols; integrity of printed
More Product Information