T-Mop™ Cleanroom Mop Covers
Supplier: MICRONOVA
QDT-10 QDT-18 T2-14 T2-18 T6-10 T6-14 T6-18 QDST-10 QDST-14 QDST-18 T6-14IR T6-18IR QDT-14 TAA-18 TAA-14 T612-18 T612-14 T1112-14 T612-18IR TAA12-14 TAA12-18 TAA-18IR T612-14IR T20-14 TMC6-14 QPT-18 QPT-14
129-0539EA 279 EUR
129-0539 129-0541 129-0556 129-0557 129-0558 129-0559 141-0877 MICRQDST-10 129-0627 129-0628 141-0876 141-0878 129-0281 141-0880 141-0879 129-0649 129-0647 MICRT1112-14 MICRT612-18IR MICRTAA12-14 MICRTAA12-18 MICRTAA-18IR MICRT612-14IR MICRT20-14 MICRTMC6-14 MICRQPT-18 MICRQPT-14
T-Mop™ Cleanroom Mop Covers
Mops
A series of rounded T-mop covers with snaps that easily roll down walls, as well as clean awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialised cleaning.
  • Low profile, rounded covered sponges
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces, and under equipment
  • Holds up to strong disinfectants
  • Autoclavable
Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester forMore Product Information