Microplate readers, multi-mode, SpectraMax® M series
Supplier: Molecular Devices
M2 M2E M3 M4 M5 M5E
MLDVM2EA 0 EUR
MLDVM2 MLDVM2E 736-0910 MLDVM4 MLDVM5 MLDVM5E
Microplate readers, multi-mode, SpectraMax® M series
Microplate Readers
All SpectraMax® M series multi-mode microplate readers measure UV and visible absorbance, and fluorescence intensity measurements on microplates and cuvettes as standard. The SpectraMax® M2 and M2e readers provide two detection modes in one platform, and offer the the added functionality of bottom-read fluorescence for cell-based assays (M2e). The SpectraMax® M3, M4, M5 and M5e readers offer a modular, upgradeable dual monochromator microplate reader platform with a wide range of high performance multi-mode capabilities ideal for life science research and drug discovery screening.