Microplate washer, MultiWash™+
Supplier: Molecular Devices
MULTIWASHPLUS
MLDVMULTIWASHPLUSEA 0 EUR
MLDVMULTIWASHPLUS
Microplate washer, MultiWash™+
Microplate Washers
The compact and portable MultiWash™+ microplate washer includes 20 preset protocols, works with both 96- and 384-well plates, and is supplied with four different wash/rinse bottles and one waste bottle for out-of-the-box use.
  • Walk-up usability
  • Automated rinse reduces clogging
  • Cross-wise aspiration reduces residual volume within each well
  • Vacuum- and pressure-free
  • Adjustable speed, volume, soak time, and aspiration speed and time
  • Up to 20 different wash protocols (Up to 8 cycles within each protocol)
The MultiWash™+ Microplate Washer features a varietyMore Product Information