Aluminium foil tapes, 431
Supplier: 3M
431-19MM 431-25MM 431-38MM 431-50MM
MMMA431-19MMEA 2140 EUR
MMMA431-19MM MMMA431-25MM MMMA431-38MM MMMA431-50MM
Aluminium foil tapes, 431
Tapes
3M™ aluminum foil tape 431 is designed for numerous applications across a broad range of industries. This tape is comprised of a dead soft aluminium foil backing and coated with a highly engineered pressure sensitive acrylic adhesive.
  • Heat and light reflective tape protects surfaces and enhances lighting efficiency
  • Thermally conductive tape reflects and dissipates radiant heat to protect plastic surfaces
  • Robust acrylic adhesive and foil backing provides long service life, both indoors and out
  • Withstands exposure to flame, weathering, moisture, and UV for a
More Product Information