Lane and safety marking vinyl tapes 471
Supplier: 3M
471BK50IW 471Y50IW 471W50IW 471GR50IW 471R50IW 471B50IW 471B9IW 471R12IW
MMMA471BK50IWEA 1150 EUR
MMMA471BK50IW MMMA471Y50IW MMMA471W50IW MMMA471GR50IW MMMA471R50IW MMMA471B50IW MMMA471B9IW MMMA471R12IW
Lane and safety marking vinyl tapes 471
Tapes
3M™ Lane and Safety Marking Tape 471 is commonly used for marking hazards and aisles, colour-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, this versatile tape resists scrapes, wear, and weathering for long-lasting use.
  • Maintains colour when exposed to abrasion, wear, and many solvents
  • Unique stretch properties enables tape to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, with excellent holding strength on many surfaces