Reusable Front Fitting Full Face Masks, FF-600F
Supplier: 3M
7100265074 7100265075 7100265236
MMMA7100265074EA 479 EUR
MMMA7100265074 MMMA7100265075 MMMA7100265236
Reusable Front Fitting Full Face Masks, FF-600F
Respirators Reusable Respirators Full Face Respirators
The FF-600F series reusable full face masks feature a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ DT series filters, these face masks are suitable for a wide range of wearers and applications.
  • Panoramic visor provides a virtually unrestricted field of vision
  • Visor is designed to minimise visual distortion
  • Soft, highly flexible, and conforming Liquid Silicone Rubber faceseal enhances user comfort
  • Broad faceseal, designed to minimise pressure points and provide an effective seal without over-tightening
More Product Information