Polyester Tape 8403
Supplier: 3M
84032566
MMMA84032566EA 4570 EUR
MMMA84032566
Polyester Tape 8403
Tapes
For anodizing, composite bonding, and splicing, use 3M™ Polyester Tape 8403. This translucent green tinted tape is coated with a silicone adhesive that bonds well on difficult surfaces, including silicone-coated papers and films. It is chemical resistant and performs under a wide range of temperatures and environments. This tape is a thicker version of 3M™ polyester tape 8402.
  • Works in demanding conditions
  • Retains flexibility in autoclave conditions in temperatures ranging from –50 to +177 °C (–60 to +350 °F)
  • Silicone adhesive provides clean removal from a
More Product Information