Removable repositionable double coated tape, 9415PC
Supplier: 3M
94151219
MMMA94151219EA 7480 EUR
MMMA94151219
Removable repositionable double coated tape, 9415PC
Tapes
3M™ removable repositionable tape 9415PC is an acrylic adhesive 400 and 1000 on a 78 lbs. polycoated kraft paper liner and polyester film carrier.
  • 25 µm polyester film carrier on a 50 µm removable, repositionable double coated tape
  • Works with lightweight materials such as paper, plastics, films, or foams
  • Remove tape without leaving residue from many films, foils and papers, due to its ultra-low tack
  • High tack/low tack
  • Medium temperature and low solvent resistance
  • Medium/low HSE and low LSE
Use 3M™ Removable Repositionable Tape 9415PC to allow removalMore Product Information