Powered air respirator headtop forehead seals for M-100/M-200 series, Versaflo™
Supplier: 3M
M154
MMMAM154EA 60.7 EUR
MMMAM154
Powered air respirator headtop forehead seals for M-100/M-200 series, Versaflo™
Respirators Respiratory Systems
The Versaflo™ Forehead Seal M-154 is a replacement forehead seal designed for use with 3M™ Versaflo™ M-100 and M-200 Series Headtops.
  • Easy to fit
  • Use with 3M™ Versaflo™ M-100 and M-200 Series Headtops
Ensure headtop seals well with this replacement 3M™ Versaflo™ Forehead Seal, M-154. This forehead seal is easy to use and designed as a spare part to replace worn or broken seals in 3M™ Versaflo™ M-100 and M-200 Series Headtops.