Surface preparation pad plus, Scotch-Brite™
Supplier: 3M
SPPP17
MMMASPPP17EA 0 EUR
MMMASPPP17
Surface preparation pad plus, Scotch-Brite™
Wipes Cleaning Kits and Pads
Scotch-Brite™ Surface Preparation Pad Plus is a premium floor stripping pad that quickly strips and prepares a variety of surfaces for finishing applications. Its design significantly improves mid-life performance compared to competitive products and is certified by Green Seal® for environmental innovation.
  • Effectively strip and prepare surfaces such as wood, vinyl tile, sheet vinyl, marble, terrazzo, and concrete floors
  • Up to two times faster finish removal than competitive pads when worn
  • Does not require the use of a backer pad (confirm pad driver
More Product Information