Oil Sorbent Boom
Supplier: 3M
T270GA
MMMAT270GAEA 987 EUR
MMMAT270GA
Oil Sorbent Boom
Spill Control Materials
3M™ Oil sorbents are an innovative range of highly absorbent, hydrophobic (water repellent), quality products.
  • Ideal for absorbing oil from areas where water may also be present
  • Ideal for placing around equipment
  • Prevent leaks or spills from spreading to adjacent areas
Will not disintegrate or sink, even when saturated with oil, and very easy to dispose of.