Safety helmets, SecureFit™ X5500 series
Supplier: 3M
X5501NVE-CE-4 X5501NVE-CE-10 X5514V-CE
MMMAX5501NVE-CE-4EA 106 EUR
MMMAX5501NVE-CE-4 MMMAX5501NVE-CE-10 MMMAX5514V-CE
Safety helmets, SecureFit™ X5500 series
Head Protection Safety Helmets
3M™ SecureFit™ safety helmet X5500 series provides secure, comfortable, and adjustable head protection for workers in a variety of situations. The helmets feature adjustable positioning and easy-turning, 4-point ratchet suspension systems with the exclusive 3M™ pressure diffusion technology. The X5500 series also feature 3M™ UVicator™ Sensors that indicate damaging levels of ultraviolet (UV) light.
  • Suspension system incorporates exclusive 3M™ pressure diffusion technology
  • 4-point ratchet suspension allows height and fit customisation with multiple levels
More Product Information