Water positive control (30 ng), HWATP10, Clean-Trace™
Supplier: 3M Food Safety
HWATP10
MMMPHWATP10EA 135 EUR
MMMPHWATP10
Water positive control (30 ng), HWATP10, Clean-Trace™
Monitoring Systems Hygiene Monitoring Systems
Designed to provide assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly.
  • Verifies the performance of reagents and instruments
  • Can be used as a quench test with 3M™ Clean-Trace™ Water ATP tests
Designed to enhance your hygiene monitoring procedures by providing assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly to ensure reliable results.