Sesamol (3,4-(Methylenedioxy)phenol)

Supplier: Molekula

10825829-25G
MOLE10825829-25GEA 102 EUR
MOLE10825829-25G
Sesamol (3,4-(Methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
Formula: C₇H₆O₃
MDL Number: MFCD00005827
CAS Number: 533-31-3
EINECS: 208-561-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR