Copper(II) trifluoromethanesulphonate

Supplier: Molekula

Synonyms: Copper(II) trifluoromethanesulphonate, Copper triflate

11742172-5G
MOLE11742172-5GEA 0 EUR
MOLE11742172-5G
Copper(II) trifluoromethanesulphonate
Copper (II) trifluoromethanesulphonate
Formula: C₂CuF₆O₆S₂
MDL Number: MFCD00077492
CAS Number: 34946-82-2
EINECS: 252-300-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR