2-Chloro-4'-fluoroacetophenone

Supplier: Molekula

11784732-25G
MOLE11784732-25GEA 0 EUR
MOLE11784732-25G
2-Chloro-4'-fluoroacetophenone
2-Chloro-4'-fluoroacetophenone
Formula: C₈H₆ClFO
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 45 °C
MDL Number: MFCD00011652
CAS Number: 456-04-2
EINECS: 207-256-4
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR