1,4-Dithiane-2,5-diol

Supplier: Molekula

11970781-1KG 11970781-100G
MOLE11970781-1KGEA 502 EUR
MOLE11970781-1KG MOLE11970781-100G
1,4-Dithiane-2,5-diol
1,4-Dithiane-2,5-diol
Formula: C₄H₈O₂S₂
MDL Number: MFCD00006659
CAS Number: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR