D-(+)-Sucrose octaacetate

Supplier: Molekula

Synonyms: D-(+)-Saccharose octaacetate, Sucrose octaacetate, D(+)-Sucroseoctaacetat, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-(((2S,3S,4R,5R)-3,4-diacetoxy-2,5-bis(acetoxymethyl)tetrahydrofuran-2-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate

12351776-1KG
MOLE12351776-1KGEA 0 EUR
MOLE12351776-1KG
D-(+)-Sucrose octaacetate
D(+)-Sucrose octaacetate
Formula: C₂₈H₃₈O₁₉
Melting Pt: 80…83 °C
Density: 1,257 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006623
CAS Number: 126-14-7
EINECS: 204-772-1
Merck Index: 12,09052

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR