1-Bromo-3-fluorobenzene

Supplier: Molekula

12409749-100G
MOLE12409749-100GEA 0 EUR
MOLE12409749-100G
1-Bromo-3-fluorobenzene
1-Bromo-3-fluorobenzene
Formula: C₆H₄BrF
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1,7081 g/cm³ (20 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00000326
CAS Number: 1073-06-9
EINECS: 214-023-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR