Cyclopentanol

Supplier: Molekula

13457158-500G
MOLE13457158-500GEA 177 EUR
MOLE13457158-500G
Cyclopentanol
Cyclopentanol
Formula: C₅H₁₀O
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –19 °C
Density: 0,949 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00001363
CAS Number: 96-41-3
EINECS: 202-504-8
UN: 2244
ADR: 3,III
Merck Index: 13,02770

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR