(S)-(+)-2-Amino-1-butanol

Supplier: Molekula
Danger

Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol

15714187-5G
MOLE15714187-5GEA 230 EUR
MOLE15714187-5G
(S)-(+)-2-Amino-1-butanol
(S)-(+)-2-Amino-1-butanol
Formula: C₄H₁₁NO
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0,947 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064418
CAS Number: 5856-62-2
EINECS: 227-475-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR