4-Cyanophenylboronic acid

Supplier: Molekula
Warning

15798945-1G
MOLE15798945-1GEA 0 EUR
MOLE15798945-1G
4-Cyanophenylboronic acid
4-Cyanophenylboronic acid
Formula: C₇H₆BNO₂
Melting Pt: > 300 °C
MDL Number: MFCD01318968
CAS Number: 126747-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR