Potassium thiocyanate

Supplier: Molekula
Warning

Synonyms: Potassium rhodanide

16005796-1KG
MOLE16005796-1KGEA 87.4 EUR
MOLE16005796-1KG
Potassium thiocyanate
Potassium thiocyanate
Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500
Melting Pt: 175
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR