12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V)

Supplier: Molekula

16255417-5G
MOLE16255417-5GEA 0 EUR
MOLE16255417-5G
12-Crown-4 (1,4,7,10-tetraoxacyclododecane, lithium ionophore V)
12-Crown-4 (1,4,7,10-Tetraoxacyclododecane, Lithium ionophore V)
Formula: C₈H₁₆O₄
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1,11 g/cm³ (20 °C)
MDL Number: MFCD00005103
CAS Number: 294-93-9
EINECS: 206-036-5
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR