3-Methoxysalicyl alcohol

Supplier: Molekula

16307918-5G
MOLE16307918-5GEA 0 EUR
MOLE16307918-5G
3-Methoxysalicyl alcohol
3-Methoxysalicyl alcohol
Formula: C₈H₁₀O₃
MDL Number: MFCD00008362
CAS Number: 4383-05-5
EINECS: 224-488-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR