3-Nitro-o-toluic acid

Supplier: Molekula

18153296-100G
MOLE18153296-100GEA 0 EUR
MOLE18153296-100G
3-Nitro-o-toluic acid
3-Nitro-o-toluic acid
Formula: C₈H₇NO₄
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00007160
CAS Number: 1975-50-4
EINECS: 217-826-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR