5-Iodosalicylic acid

Supplier: Molekula
Warning

Synonyms: 2-Hydroxy-5-iodobenzoic acid

21549902-100G
MOLE21549902-100GEA 0 EUR
MOLE21549902-100G
5-Iodosalicylic acid
5-Iodosalicylic acid
Formula: C₇H₅IO₃
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 197…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00002458
CAS Number: 119-30-2
EINECS: 204-313-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR