Barium sulphate

Supplier: Molekula

21615331-1KG
MOLE21615331-1KGEA 87.7 EUR
MOLE21615331-1KG
Barium sulphate
Barium sulphate
Formula: BaSO₄
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 1345 °C
Density: 4,5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR