Sodium cyanotrihydroborate

Supplier: Molekula
Danger

Synonyms: Sodium cyanoborohydride

23955696-25G
MOLE23955696-25GEA 88 EUR
MOLE23955696-25G
Sodium cyanotrihydroborate
Sodium cyanotrihydroborate
Formula: CH₃BNNa
Melting Pt: 241…243 °C
Density: 1,11…1.12 g/cm³ (28 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3,II
Merck Index: 13,08680

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR