Aluminium chloride hexahydrate

Supplier: Molekula
Warning

Synonyms: Aluminum chloride hexahydrate

26726139-100G
MOLE26726139-100GEA 0 EUR
MOLE26726139-100G
Aluminium chloride hexahydrate
Aluminium chloride hexahydrate
Formula: AlCl₃·6H₂O
Melting Pt: ∼100 °C
Density: 2,39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR