Quinine sulphate dihydrate

Supplier: Molekula

29117911-25G
MOLE29117911-25GEA 146 EUR
MOLE29117911-25G
Quinine sulphate dihydrate
(-)-Quinine sulphate dihydrate
Formula: C₄₀H₅₀N₄O₈S·2H₂O
Melting Pt: 219 °C
Density: 1,36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150790
CAS Number: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR